1,12-Dodecanedioic acid 99%

Warning

Synonyms: Dodecanedioic acid1,10-Decanedicarboxylic acid

AC11188-5000 AC11188-1000 AC11188-0050
200015-466EA 89.3 USD
200015-466 200027-530 103508-748
1,12-Dodecanedioic acid 99%
1,12-Dodecanedioic acid
Formula: C₁₂H₂₂O₄
MW: 230.3 g/mol
Boiling Pt: 245 °C (10 mmHg)
Melting Pt: 125…130 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002735
CAS Number: 693-23-2
EINECS: 211-746-3

Order Now

Specification Test Results

Appearance White Powder or pellets
Infrared spectrum Conforms
Melting point 125 to 130 °C
Titration with NaOH ≥98.5 %
Viscosity 30.5 to 36.5
pH 3.0 to 3.8 (1 % solution)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR