Bromocyclopropane 99%

Danger

Synonyms: Cyclopropyl bromide

AC11164-0250 AC11164-0050
200018-636EA 164 USD
200018-636 200011-868
Bromocyclopropane 99%
Bromocyclopropane
Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 67…69 °C (1013 hPa)
Density: 1.51 g/cm³
Flash Pt: –7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001271
CAS Number: 4333-56-6
EINECS: 224-375-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20 °C, 589 nm)
GC ≥98.5 %
Absorption Ratio P-15/P+15 0.93 to 0.98
BSC Certification Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR