Bromocyclopentane 98%

Warning

Synonyms: Cyclopentyl bromide

AC11160-2500 AC11160-0500 AC11160-0010
200011-850EA 27.4 USD
200011-850 103524-838 200006-166
Bromocyclopentane 98%
Bromocyclopentane
Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 137…139 °C (760 mmHg)
Density: 1.39 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00001359
CAS Number: 137-43-9
EINECS: 205-294-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4881 to 1.4901 (20 °C, 589 nm)
GC ≥97.5 %
UV (5 % in water)
UV at 210 nm A: ≤0.1
UV at 220 nm A: ≤0.07
UV at 230 nm A: ≤0.06
UV at 250 nm A: ≤0.03
UV at 260 nm A: ≤0.03
Titration after Ion exchange 98.5 to 101.5 % (on dry substance)
Chloride (Cl) ≤1000 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR