Cyclooctylamine 97%

Danger

Synonyms: CyclooctanamineAminocyclooctane

AC11146-1000 AC11146-0050 AC11146-0250
200043-036EA 33 USD
200043-036 200024-862 103530-478
Cyclooctylamine 97%
Cyclooctylamine
Formula: C₈H₁₇N
MW: 127.23 g/mol
Boiling Pt: 190 °C (760 mmHg)
Melting Pt: –48 °C
Density: 0.92 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00001748
CAS Number: 5452-37-9
EINECS: 226-694-7
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4820 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR