Cyclooctanone 98%

Danger

AC11144-0250 AC11144-1000 AC11144-5000
200018-622EA 88.3 USD
200018-622 103530-474 103530-476
Cyclooctanone 98%
Cyclooctanone
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 195…197 °C (760 mmHg)
Melting Pt: 39…44 °C
Density: 0.95 g/cm³
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00001754
CAS Number: 502-49-8
EINECS: 207-940-2
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to white Low melting crystalline solid
Infrared spectrum Conforms
GC ≥97.5 %
Titration with TBAH ≥97.5 %
GC ≥99.50 %
Color scale ≤50 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR