Cyclohexylamine ≥99%

Supplier: ACROS ORGANICS MS

Danger

Synonyms: Aminocyclohexane

AC11128-2500 AC11128-0025 AC11128-0010 AC11128-0100
200000-262EA 23.63 USD
200000-262 200006-162 200000-260 103524-828
Cyclohexylamine ≥99%
Cyclohexylamine
Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 133…134 °C (760 mmHg)
Melting Pt: –17 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00001486
CAS Number: 108-91-8
EINECS: 203-629-0
UN: 2357
ADR: 8,II
Merck Index: 13,02758

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4600 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR