Cyclohexanone oxime 97%

Warning

AC11120-5000 AC11120-1000
200000-258EA 56.2 USD
200000-258 200030-798
Cyclohexanone oxime 97%
Cyclohexanone oxime
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: 88…92 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001660
CAS Number: 100-64-1
EINECS: 202-874-0
UN: 1325

Order Now

Specification Test Results

Appearance Off-white to beige to light brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 88 to 92 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR