1,3-Cyclohexanedione 97%

Warning

Synonyms: Cyclohexane-1,3-dione

AC11117-5000 AC11117-0025 AC11117-1000
200056-762EA 182 USD
200056-762 103508-730 200056-760
1,3-Cyclohexanedione 97%
1,3-Cyclohexanedione
Formula: C₆H₈O₂
MW: 112.13 g/mol
Melting Pt: >101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001585
CAS Number: 504-02-9
EINECS: 207-980-0
Merck Index: 13,03200

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point ≥101 °C
Titration Argentometric ≤1 % Sodium chloride
Water ≤0.5 %
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR