1,2-Cyclohexanedione 98%

Warning

AC11116-2500 AC11116-0500 AC11116-0100
103508-728EA 1577 USD
103508-728 200008-418 200021-572
1,2-Cyclohexanedione 98%
1,2-Cyclohexanedione
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 193…195 °C (760 mmHg)
Melting Pt: 35…38 °C
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001648
CAS Number: 765-87-7
EINECS: 212-155-3

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5180 to 1.5200 (20 °C, 589 nm)
GC ≥97.5 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.005 %
Magnesium (Mg) ≤0.01 %
Manganese (Mn) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR