Cycloheptanone 98+%

Warning

Synonyms: Cycloheptan-1-oneSuberon

AC11104-0250 AC11104-5000 AC11104-1000
200011-818EA 135 USD
200011-818 103530-024 103530-026
Cycloheptanone 98+%
Cycloheptanone
Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 179 °C (760 mmHg)
Melting Pt: –21 °C
Density: 0.95 g/cm³
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00004159
CAS Number: 502-42-1
EINECS: 207-937-6
UN: 1224
ADR: 3,III
Merck Index: 13,02751

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4595 to 1.4635 (20 °C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR