1,1-Cyclobutanedicarboxylic acid 99%

Danger

Synonyms: Cyclobutane-1,1-dicarboxylic acid

AC11092-0250 AC11092-0050
200027-536EA 43.7 USD
200027-536 200039-910
1,1-Cyclobutanedicarboxylic acid 99%
1,1-Cyclobutanedicarboxylic acid
Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 663 °C (1013 hPa)
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00001325
CAS Number: 5445-51-2
EINECS: 226-651-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 157 to 161 °C
Water ≤0.1 % (K.F.)
Loss on drying ≤0.1 % (50 °C)
GC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %
Clarity of solution (5 w/v% aq. soln.) clear

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR