Cyclobutanecarboxylic acid 98%

Warning

AC11090-0050 AC11090-0250
200011-808EA 78.2 USD
200011-808 200042-976
Cyclobutanecarboxylic acid 98%
Cyclobutanecarboxylic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 195 °C (760 mmHg)
Melting Pt: –20 °C
Density: 1.04 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00001323
CAS Number: 3721-95-7
EINECS: 223-072-7
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4450 (20 °C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR