Isonicotinonitrile 98%

Warning

Synonyms: 4-Pyridinecarbonitrile4-CyanopyridinePyridine-4-carbonitrile

AC11087-1000 AC11087-5000 AC11087-0250 AC11087-0050
200041-966EA 17.3 USD
200041-966 200029-776 103508-724 200010-380
Isonicotinonitrile 98%
Isonicotinonitrile
Formula: C₆H₄N₂
MW: 104.11 g/mol
Boiling Pt: 196 °C (760 mmHg)
Melting Pt: 77…81 °C
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00006417
CAS Number: 100-48-1
EINECS: 202-856-2

Order Now

Specification Test Results

Appearance White to yellow to beige to light brown Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR