Nicotinonitrile 98%

Warning

Synonyms: Pyridine-3-carbonitrile3-Cyanopyridine3-Pyridinecarbonitrile

AC11086-0025 AC11086-1000 AC11086-0050 AC11086-5000
200029-266EA 27.5 USD
200029-266 200009-902 200041-530 200041-528
Nicotinonitrile 98%
Nicotinonitrile
Formula: C₆H₄N₂
MW: 104.11 g/mol
Boiling Pt: 206.2 °C (1013 hPa)
Melting Pt: 47…51 °C
Density: 1.159 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00087130
CAS Number: 100-54-9
EINECS: 202-863-0

Order Now

Specification Test Results

Appearance Colorless to white to yellow Crystals or crystalline mass and/or chunks
Infrared spectrum Conforms
Melting point 47 to 51 °C
Water ≤0.2 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR