Pyridine-2-carbonitrile 99%

Warning

Synonyms: 2-PyridinecarbonitrilePicolinonitrile2-Cyanopyridine

AC11085-5000 AC11085-0050 AC11085-1000
200041-000EA 21.8 USD
200041-000 200016-540 200009-346
Pyridine-2-carbonitrile 99%
Pyridine-2-carbonitrile
Formula: C₆H₄N₂
MW: 104.11 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 26…28 °C
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00006218
CAS Number: 100-70-9
EINECS: 202-880-3
UN: 3439

Order Now

Specification Test Results

Appearance White to brown Fused solid or liquid
Infrared spectrum Conforms
Melting point 25 to 31 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR