Cyanogen bromide 97%

Danger

AC11078-1000 AC11078-0250
200000-244EA 136 USD
200000-244 200000-242
Cyanogen bromide 97%
Cyanogen bromide
Formula: CBrN
MW: 105.92 g/mol
Boiling Pt: 61.4 °C (1013 hPa)
Melting Pt: 49…54 °C
Flash Pt: >65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011597
CAS Number: 506-68-3
EINECS: 208-051-2
UN: 1889
ADR: 6.1,I
Merck Index: 12,02763

Order Now

Specification Test Results

Appearance White Crystals
Titration Argentometric 96.0 to 103.0 %
Identification Bromides : positive
Clarity of solution (Aqueous) Passes test
Clarity of alcohol solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR