α-Chloro-p-xylene 98%

Danger

Synonyms: alpha-Chloro-p-xylene4-Methylbenzyl chloride

AC11017-1000 AC11017-5000
200042-524EA 48.8 USD
200042-524 200018-094
α-Chloro-p-xylene 98%
α-Chloro-p-xylene
Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.06 g/cm³
Flash Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000919
CAS Number: 104-82-5
EINECS: 203-241-1
UN: 3265
ADR: 8,II
Merck Index: 12,10224

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5330 to 1.5370 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR