1-Chloropropane 99%

Danger

AC10995-1000 AC10995-0050 AC10995-5000
200015-774EA 36.6 USD
200015-774 200004-718 103524-782
1-Chloropropane 99%
1-Chloropropane
Formula: C₃H₇Cl
MW: 78.54 g/mol
Boiling Pt: 46…47 °C (760 mmHg)
Melting Pt: –123 °C
Density: 0.892 g/cm³
Flash Pt: –18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000995
CAS Number: 540-54-5
EINECS: 208-749-7
UN: 1278
ADR: 3,II
Merck Index: 12,08032

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3870 to 1.3900 (20 °C, 589 nm)
GC ≥98.5 %
Iron (Fe) ≤50 ppm
Mercurous Mercury (as Hg) ≤0.15 %
Residue after reduction ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR