1-(Chloromethyl)naphthalene 95%

Danger

AC10945-5000 AC10945-1000 AC10945-0050
200015-438EA 44.9 USD
200015-438 200039-858 200027-506
1-(Chloromethyl)naphthalene 95%
1-(Chloromethyl)naphthalene
Formula: C₁₁H₉Cl
MW: 176.65 g/mol
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 31…31 °C
Density: 1.17 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004042
CAS Number: 86-52-2
EINECS: 201-678-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.6350 to 1.6390 (20 °C, 589 nm)
HPLC ≥94 %
HPLC Remainder 2-chloromethylnaphtalene
Acid value ≤35 mg KOH/g
Peroxide value ≤4 meq O2/kg
UV at 280 nm A: ≤0.1
Residue after ign. (sulfated) ≤0.3 %
Specific optical rotation +102° to +106° (20 °C, 589 nm) (10% in water)
Specific optical rotation (after 24 h)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR