4-Chloro-3,5-xylenol 99%

Warning

Synonyms: Chloroxylenol4-Chloro-3,5-dimethylphenol2-Chloro-5-hydroxy-3-xylene

AC10906-5000 AC10906-0025 AC10906-1000
103524-778EA 284 USD
103524-778 200029-702 200023-688
4-Chloro-3,5-xylenol 99%
4-Chloro-3,5-xylenol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 246 °C (760 mmHg)
Melting Pt: 113…116 °C
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00002324
CAS Number: 88-04-0
EINECS: 201-793-8
UN: 3077
ADR: 9,III
Merck Index: 13,02194

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 113 to 116 °C
GC ≥98.5 %
Stabilizer ca. 500 ppm MEHQ

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR