1-Chlorodecane 97-99%

Warning

AC10898-2500 AC10898-0010
200015-762EA 113 USD
200015-762 103530-442
1-Chlorodecane 97-99%
1-Chlorodecane
Formula: C₁₀H₂₁Cl
MW: 176.73 g/mol
Boiling Pt: 223 °C (760 mmHg)
Melting Pt: –34 °C
Density: 0.86 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00000956
CAS Number: 1002-69-3
EINECS: 213-691-0
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4362 to 1.4382 (20 °C, 589 nm)
GC ≥97 %
Residue after evaporation ≤0.003 %
Carbonyl (as HCHO) ≤0.1 %
Acids and esters ≤0.2 % (as Amyl acetate)
Titrable acid ≤0.002 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR