2-Chlorobutane 99+%

Danger

AC10883-0010 AC10883-2500
200009-314EA 176.54 USD
200009-314 200000-212
2-Chlorobutane 99+%
2-Chlorobutane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 68 °C (1013 hPa)
Melting Pt: –140 °C
Density: 0.87 g/cm³
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00000871
CAS Number: 78-86-4
EINECS: 201-151-7
UN: 1127
ADR: 3,II
Merck Index: 13,01560

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3980 (20 °C, 589 nm)
GC ≥99.0 %
UV at 219 nm A: ≤1
UV at 230 nm A: ≤0.5
UV at 245 nm A: ≤0.1
UV at 250 nm A: ≤0.05
UV at 260 nm A: ≤0.03
UV at 330 nm A: ≤0.01
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR