(4-Chlorophenyl)acetonitrile 98+%

Danger

Synonyms: 4-Chlorobenzyl cyanide

AC10881-5000 AC10881-2500 AC10881-0050 AC10881-1000
200023-720EA 26.9 USD
200023-720 200029-744 200035-826 200041-928
(4-Chlorophenyl)acetonitrile 98+%
(4-Chlorophenyl)acetonitrile
Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 29…30 °C
Flash Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00001918
CAS Number: 140-53-4
EINECS: 205-418-9
UN: 3439
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5415 to 1.5435 (20 °C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR