4-Chloro-α,α,α-trifluorotoluene 98%

Warning

Synonyms: 4-Chlorobenzotrifluoride1-Chloro-4-(trifluoromethyl)benzene

AC10873-2500 AC10873-0010 AC10873-0250
200017-476EA 22.8 USD
200017-476 200010-340 103530-434
4-Chloro-α,α,α-trifluorotoluene 98%
4-Chloro-α,α,α-trifluorotoluene
Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Boiling Pt: 136…138 °C (760 mmHg)
Melting Pt: –36 °C
Density: 1.35 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00000627
CAS Number: 98-56-6
EINECS: 202-681-1
UN: 2234
ADR: 3,III
Merck Index: 13,02145

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4453 to 1.4473 (20 °C, 589 nm)
GC ≥97.5 %
Stabilizer copper
Acidity (CH3COOH) ≤0.002 %
Peroxides ≤0.005 % (as H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR