4-Chlorobenzophenone ≥99%

Synonyms: p-Chlorobenzophenone

AC10869-1000
200023-716EA 34.5 USD
200023-716
4-Chlorobenzophenone ≥99%
4-Chlorobenzophenone
Formula: C₁₃H₉ClO
MW: 216.67 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00000622
CAS Number: 134-85-0
EINECS: 205-160-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 75 to 77 °C
Loss on drying ≤0.5 %
GC ≥98.5 %
Peroxides ≤0.002 % (as H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR