2-Chlorobenzonitrile 99%
|
Warning
|
Synonyms:
o-Chlorobenzonitrile
AC10867-0010
AC10867-2500
200028-644EA
68.3
USD
200028-644
200040-942
2-Chlorobenzonitrile 99%
2-Chlorobenzonitrile
Formula:
C₇H₄ClN MW: 137.57 g/mol Boiling Pt: 232 °C (1013 hPa) Melting Pt: 43…46 °C Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001779 CAS Number: 873-32-5 EINECS: 212-836-5 ADR: 6.1,III |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals and/or chunks or solid |
Infrared spectrum | Conforms |
Melting point | 43 to 46 °C |
Water | ≤0.5 % |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...