3-Chlorobenzaldehyde 99%

Warning

AC10861-1000 AC10861-5000 AC10861-0250
200017-004EA 189 USD
200017-004 103465-358 200009-882
3-Chlorobenzaldehyde 99%
3-Chlorobenzaldehyde
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 213…215 °C (760 mmHg)
Melting Pt: 17…18 °C
Density: 1.241 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003350
CAS Number: 587-04-2
EINECS: 209-596-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5650 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR