3-Chloroaniline 99%

Danger

AC10858-1000 AC10858-5000 AC10858-0050
200009-878EA 45.5 USD
200009-878 200017-002 200023-238
3-Chloroaniline 99%
3-Chloroaniline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 230.5 °C (1013 hPa)
Melting Pt: –11 °C
Density: 1.2 g/cm³
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00007765
CAS Number: 108-42-9
EINECS: 203-581-0
UN: 2019
ADR: 6.1,II
Merck Index: 13,02136

Order Now

Specification Test Results

Appearance Clear light yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5927 to 1.5947 (20 °C, 589 nm)
GC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR