Butyrylthiocholine iodide 98%

Warning

Synonyms: 2-(Butyrylthio)-N,N,N-trimethylethan-1-aminium iodide(2-(Butyrylthio)ethyl)trimethylammonium iodide(2-Butyrylthioethyl)trimethylammonium iodide(Propylcarbonylthioethyl)trimethylammonium iodide

AC10815-0250 AC10815-1000
200038-762EA 125 USD
200038-762 103508-942
Butyrylthiocholine iodide 98%
Butyrylthiocholine iodide
Formula: H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I
MW: 317.23 g/mol
Melting Pt: 172…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011845
CAS Number: 1866-16-6
EINECS: 217-475-7

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 172 to 176 °C
Titration Argentometric ≥97.5 %
UV at 210 nm A: ≤0.4
UV at 220 nm A: ≤0.15
UV at 230 nm A: ≤0.05
UV at 240 nm A: ≤0.01
UV at 250 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR