Butyronitrile 99%

Danger

Synonyms: n-Butyronitrile

AC10814-5000 AC10814-0025 AC10814-1000
200055-990EA 136 USD
200055-990 103524-760 200055-988
Butyronitrile 99%
Butyronitrile
Formula: C₄H₇N
MW: 69.11 g/mol
Boiling Pt: 115…117 °C (760 mmHg)
Melting Pt: –112 °C
Density: 0.79 g/cm³
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00001968
CAS Number: 109-74-0
EINECS: 203-700-6
UN: 2411
ADR: 3,II
Merck Index: 13,01597

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3850 (20 °C, 589 nm)
GC ≥98.5 %
Color scale ≤25 APHA
UV at 250 nm A: ≤0.005
Color scale ≤5 APHA
Residue after evaporation ≤2 ppm
Acidity ≤0.0002 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR