2-Butyne-1,4-diol 99%

Danger

Synonyms: But-2-yne-1,4-diol1,4-Dihydroxy-2-butyne

AC10808-0010 AC10808-2500 AC10808-0025
200016-436EA 39.95 USD
200016-436 103524-754 200005-008
2-Butyne-1,4-diol 99%
2-Butyne-1,4-diol
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 238 °C (760 mmHg)
Melting Pt: 54…58 °C
Density: 1.2 g/cm³
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00002915
CAS Number: 110-65-6
EINECS: 203-788-6
UN: 2716
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to beige Crystals or flakes
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR