4-tert-Butylphenol 97%

Danger

AC10800-0050 AC10800-0010 AC10800-0025
200005-542EA 25.5 USD
200005-542 200005-546 200017-712
4-tert-Butylphenol 97%
4-tert-Butylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 96…100 °C
Density: 0.9 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002367
CAS Number: 98-54-4
EINECS: 202-679-0
UN: 3077
ADR: 9,III
Merck Index: 13,01584

Order Now

Specification Test Results

Appearance White to light beige Flakes or pastilles
Infrared spectrum Conforms
Melting point 96 to 100 °C
Water ≤0.5 % (K.F.)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR