tert-Butylbenzene 99%

Warning

AC10787-5000 AC10787-0025 AC10787-0010
200014-658EA 80.9 USD
200014-658 200014-654 103530-274
tert-Butylbenzene 99%
tert-Butylbenzene
Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 169 °C (760 mmHg)
Melting Pt: –58 °C
Density: 0.86 g/cm³
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00008816
CAS Number: 98-06-6
EINECS: 202-632-4
UN: 2709
ADR: 3,III
Merck Index: 13,01550

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Water ≤0.5 % (K.F.)
GC ≥99.5 %
Color scale ≤10 APHA
Residue after ignition ≤0.005 %
Substance darkened by H2SO4 Passes test
Sulfate (SO4) ≤10 ppm
Chlorinated cmpds (as Cl) ≤0.003 %
Heavy metals (as Pb) ≤2 ppm
Impurity (Acrolein and glucose) Passes test
Fatty acid esters ≤0.05 % (as Butyric acid)
Neutrality Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR