(±)-1,3-Butanediol 99%, Extra Pure

Synonyms: Butane-1,3-diol(±)-1,3-Dihydroxy butane(±)-1,3-Butylene glycol

AC10762-0100 AC10762-0025 AC10762-2500 AC10762-0010
200004-632EA 166 USD
200004-632 200000-156 103508-926 103508-928
(±)-1,3-Butanediol 99%, Extra Pure
(±)-1,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 203…204 °C (760 mmHg)
Melting Pt: –54 °C
Density: 1.01 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00004554
CAS Number: 107-88-0
EINECS: 203-529-7
Merck Index: 13,01566

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4410 (20 °C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %
Color scale ≤10 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR