3-Bromo-o-xylene 99%

Warning

Synonyms: 1-Bromo-2,3-dimethylbenzene

AC10758-0050
200009-824EA 37.5 USD
200009-824
3-Bromo-o-xylene 99%
3-Bromo-o-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 214 °C (1013 hPa)
Density: 1.36 g/cm³
Flash Pt: 80 °C
MDL Number: MFCD00000069
CAS Number: 576-23-8
EINECS: 209-398-2
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5590 to 1.5610 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR