Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-p-xylene ≥98%

Supplier: ACROS ORGANICS MS

Danger
Synonyms: alpha-Bromo-p-xylene4-(Bromomethyl)toluene4-Methylbenzyl bromide

AC10757-0050 AC10757-0250 AC10757-1000
200030-366EA 26.01 USD InStock
200030-366 200011-100 103524-726
α-Bromo-p-xylene ≥98%
α-Bromo-p-xylene


Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 218…220 °C (760 mmHg)
Melting Pt: 33…37 °C
Density: 1.324 g/cm³ (20 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00000181
CAS Number: 104-81-4
EINECS: 203-240-6
UN: 3417
ADR: 6.1,II
Merck Index: 12,10223
Order Now
Specification Test Results
Appearance White to light yellow Crystalline mass
Infrared spectrum Conforms
Melting point 33°C to 37°C
GC ≥97.5 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty