α-Bromo-m-xylene 96%

Danger

Synonyms: 3-Methylbenzyl bromide

AC10756-1000 AC10756-0250
200011-094EA 74.09 USD
200011-094 103530-256
α-Bromo-m-xylene 96%
α-Bromo-m-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 185 °C (340 mmHg)
Melting Pt: 18 °C
Density: 1.37 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000177
CAS Number: 620-13-3
EINECS: 210-625-2
UN: 1701
ADR: 6.1,II
Merck Index: 12,10223

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5640 to 1.5680 (20 °C, 589 nm)
GC ≥95 %
Water ≤1 % (K.F.)
Density (20 °C) 0.869 to 0.874
GC ≥60 % (oleic acid)
Color scale ≤70 APHA
Stabilizer 80 to 120 ppm BHA content
Acid value ≤0.5 mg KOH/g
Saponification value 177 to 188 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR