(3-Bromopropyl)benzene 98%

Warning

Synonyms: 1-Bromo-3-phenylpropane

AC10729-1000 AC10729-5000 AC10729-0050
200015-714EA 26.7 USD
200015-714 103508-922 200008-598
(3-Bromopropyl)benzene 98%
(3-Bromopropyl)benzene
Formula: BrH₂CCH₂CH₂(C₆H₅)
MW: 199.09 g/mol
Boiling Pt: 219…220 °C (760 mmHg)
Density: 1.31 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00000257
CAS Number: 637-59-2
EINECS: 211-294-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR