4-Bromophenylboronic acid 98%

Warning

Synonyms: Dihydroxy(4-bromophenyl)borane(4-Bromophenyl)boronic acid(4-Bromophenyl)metaboric acid4-Bromobenzeneboronic acid

AC10728-0050 AC10728-0250
200017-420EA 76 USD
200017-420 103508-920
4-Bromophenylboronic acid 98%
4-Bromophenylboronic acid
Formula: C₆H₆BBrO₂
MW: 200.83 g/mol
Melting Pt: 287…292 °C
Storage Temperature: Ambient
MDL Number: MFCD00002104
CAS Number: 5467-74-3
EINECS: 226-779-9

Order Now

Specification Test Results

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR