(±)-2-Bromopentane 90% tech.

Danger

Synonyms: (±)-2-Pentyl bromide

AC10723-1000 AC10723-5000
200028-578EA 77.6 USD
200028-578 200009-244
(±)-2-Bromopentane 90% tech.
(±)-2-Bromopentane
Formula: H₃CBrCH(CH₂)₂CH₃
MW: 151.05 g/mol
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: –95.5 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00000160
CAS Number: 107-81-3
EINECS: 203-521-3
UN: 2343
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4393 to 1.4413 (20 °C, 589 nm)
GC ≥88 % Typical
GC ≤12 % (3-Bromopentane) Typical
Color scale ≤100 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR