4-Bromoisoquinoline 98%

Warning

AC10709-0010 AC10709-0100
200010-280EA 70.4 USD
200010-280 200023-656
4-Bromoisoquinoline 98%
4-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 39…43 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006904
CAS Number: 1532-97-4
EINECS: 216-244-8
UN: 2811

Order Now

Specification Test Results

Appearance White to light beige Crystalline solid
Infrared spectrum Conforms
Melting point 39 to 43 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR