1-Bromo-2-fluorobenzene 99%

Warning

Synonyms: 2-Bromofluorobenzene

AC10699-0500 AC10699-0100
200022-492EA 153 USD
200022-492 200016-412
1-Bromo-2-fluorobenzene 99%
1-Bromo-2-fluorobenzene
Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 155 °C (1013 hPa)
Density: 1.6 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00000282
CAS Number: 1072-85-1
EINECS: 214-018-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5327 to 1.5347 (20 °C, 589 nm)
GC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR