4-Bromo-3,5-xylenol 99%

Warning

AC10687-0250 AC10687-1000 AC10687-0100 AC10687-5000
200010-248EA 154 USD
200010-248 200029-638 103508-896 103508-898
4-Bromo-3,5-xylenol 99%
4-Bromo-3,5-xylenol
Formula: C₈H₉BrO
MW: 201.06 g/mol
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002315
CAS Number: 7463-51-6
EINECS: 231-255-8

Order Now

Specification Test Results

Appearance Colorless to light pink Powder or crystals
Infrared spectrum Conforms
Melting point 113 to 116 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR