1-Bromo-3-chloropropane 99%

Danger

AC10686-0025 AC10686-0010 AC10686-2500
200008-592EA 163.69 USD
200008-592 200004-700 103524-706
1-Bromo-3-chloropropane 99%
1-Bromo-3-chloropropane
Formula: BrH₂CCH₂CH₂Cl
MW: 157.44 g/mol
Boiling Pt: 142…145 °C (760 mmHg)
Melting Pt: –59 °C
Density: 1.59 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00000998
CAS Number: 109-70-6
EINECS: 203-697-1
UN: 2688
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4880 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR