1-Bromo-4-chlorobenzene ≥99%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: 4-Bromochlorobenzene

AC10683-0010 AC10683-0500 AC10683-1000 AC10683-5000
200029-644EA 102.6 USD InStock
200029-644 103530-210 103530-212 200029-646
1-Bromo-4-chlorobenzene ≥99%
1-Bromo-4-chlorobenzene
Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 196 °C (760 mmHg)
Melting Pt: 64…67 °C
Density: 1.576 g/cm³ (71 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000600
CAS Number: 106-39-8
EINECS: 203-392-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 64°C to 67°C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR