1-Bromobutane 99%

Danger

Synonyms: Butyl bromide

AC10677-0100 AC10677-0500 AC10677-2500 AC10677-0010 AC10677-0025
103524-702EA 22.4 USD
103524-702 103524-700 200000-126 103524-698 200000-124
1-Bromobutane 99%
1-Bromobutane
Formula: BrH₂C(CH₂)₂CH₃
MW: 137.02 g/mol
Boiling Pt: 100…104 °C (760 mmHg)
Melting Pt: –112 °C
Density: 1.27 g/cm³
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000260
CAS Number: 109-65-9
EINECS: 203-691-9
UN: 1126
ADR: 3,II
Merck Index: 13,01552

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4384 to 1.4404 (20 °C, 589 nm)
GC ≥98.5 %
Gravimetry ≥99.0 %
Suitability (for Ni determination) Passes test
Insoluble matter ≤0.05 % (in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR