4-Bromobenzoyl chloride 98%

Danger

Synonyms: p-Bromobenzoyl chloride

AC10676-0250 AC10676-1000 AC10676-0100
200000-122EA 77.2 USD
200000-122 103524-696 200010-260
4-Bromobenzoyl chloride 98%
4-Bromobenzoyl chloride
Formula: C₇H₄BrClO
MW: 219.46 g/mol
Boiling Pt: 246 °C (760 mmHg)
Melting Pt: 36…41 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000683
CAS Number: 586-75-4
EINECS: 209-580-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light brown Low melting solid
Infrared spectrum Conforms
Melting point 36 to 41 °C
Titration RCOCl ≥97.5 %
UV at 275 nm A: ≤0.3
UV at 295 nm A: ≤0.1
UV at 310 nm A: ≤0.05
UV at 340 nm A: ≤0.01
UV at 400 nm A: ≤0.01
Color scale ≤15 APHA
Residue after evaporation ≤0.005 %
Titrable acid ≤0.0005 meq/g
Titrable base ≤0.003 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR