4-Bromobenzonitrile 98%

Warning

Synonyms: p-Bromobenzonitrile

AC10671-0500 AC10671-0100
103530-204EA 150 USD
103530-204 200005-394
4-Bromobenzonitrile 98%
4-Bromobenzonitrile
Formula: C₇H₄BrN
MW: 182.02 g/mol
Boiling Pt: 235…237 °C (760 mmHg)
Melting Pt: 111…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00001811
CAS Number: 623-00-7
EINECS: 210-764-9
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder or flakes
Infrared spectrum Conforms
Melting point 111 to 114 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR