3-Bromobenzonitrile 99%

Warning

Synonyms: m-Bromobenzonitrile

AC10670-0100 AC10670-0025 AC10670-1000
200016-966EA 73 USD
200016-966 103530-200 103530-202
3-Bromobenzonitrile 99%
3-Bromobenzonitrile
Formula: C₇H₄BrN
MW: 182.02 g/mol
Boiling Pt: 225 °C (760 mmHg)
Melting Pt: 37…41 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001796
CAS Number: 6952-59-6
EINECS: 230-127-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow to beige Crystalline powder or crystals or crystalline mass
Infrared spectrum Conforms
Melting point 37 to 41 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR