9-Bromoanthracene 96%

Warning

AC10664-0050 AC10664-0250
200010-872EA 59.9 USD
200010-872 200010-870
9-Bromoanthracene 96%
9-Bromoanthracene
Formula: C₁₄H₉Br
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3
EINECS: 216-359-3

Order Now

Specification Test Results

Appearance Yellow to green to brown Powder or crystals or needles
Infrared spectrum Conforms
GC ≥95.0 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR